Anti-HTR2A Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy7®)
BIRBORB130692-100
: Biorbyt
- Antibody type:Primary
- Antigen name:5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled
- Antigen symbol:HTR2A
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)/Cy7®
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:52 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: HTR2A (5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat