Anti-Ribonucleoside-diphosphate reductase, large subunit Rabbit Polyclonal Antibody
BIRBORB109562-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Ribonucleoside-diphosphate reductase, large subunit
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide derived from the human RRM1 protein
- Purification:purified by antigen immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Ribonucleoside-diphosphate reductase, large subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human