Anti-GAPDH Rabbit polyclonal antibody
BIRBORB109439-200
: Biorbyt
- Antibody type:Primary
- Antigen name:Glyceraldehyde-3-phosphate dehydrogenase
- Antigen symbol:GAPDH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide derived from the human GAPDH alpha protein.
- Purification:purified by ammonium sulfate precipitation
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human