Anti-PTK2, FAK1 Rabbit Polyclonal Antibody
Catalog # BIRBORB109345-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:PTK2, FAK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:C-terminal region of human
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PTK2, FAK1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human