Anti-LKB1/STK11 Serine/Threonine Kinase Rabbit Polyclonal Antibody
BIRBORB108932-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Serine/Threonine Kinase
- Antigen symbol:LKB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:STK11
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide derived from the human LKB1/STK11 serine/threonine kinase protein.
- Purification:purified by ammonium sulfate precipitation
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: LKB1/STK11 Serine/Threonine Kinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human