Order Entry
Export
ContactUsLinkComponent
Anti-MEK1/2 Rabbit Polyclonal Antibody
Anti-MEK1/2 Rabbit Polyclonal Antibody
Catalog # BIRBORB108914-10
Supplier:  Biorbyt
undefined
Anti-MEK1/2 Rabbit Polyclonal Antibody
Catalog # BIRBORB108914-10
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    MEK1/2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic phosphopeptide corresponding to amino acid residues surrounding the phospho Ser218/222 of human MEK1/2
  • Purification:
    purified by affinity chromatography
  • Size:
    10 Tests
  • Pk:
    10

Specifications

About this item

Type: Primary
Antigen: MEK1/2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: