Anti-IgG4 Mouse Monoclonal Antibody
Catalog # BIRBORB108346-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:IgG4
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:1,0 mg/ml (OD280, E(0,1%)=1,4), Titer: 1:100,000 (Indirect ELISA)
- Shipping temperature:Ice
- Immunogen:IgG4 from human serum.
- Purification:purified by affinity chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: IgG4
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: