Anti-Estradiol-17-beta-6 Rabbit Polyclonal Antibody
Catalog # BIRBORB108205-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Estradiol-17-beta-6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Estradiol-6-CMO
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Estradiol-17-beta-6
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: