Anti-MEk1/2 Rabbit Polyclonal Antibody
: Biorbyt
Western Blot: 1:500-1:2000
Type: Primary
Antigen: MEk1/2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- BIRBORB106478-100
- BIRBORB106478-20
- Antigen symbol:
- MEK1/2
- MEK1/2
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Immunogen:
- A synthesized peptide derived from human MEK1/2
- A synthesized peptide derived from human MEK1/2
- Molecular weight:
- 45 kDa
- 45 kDa
- Purification:
- purified by immunoaffinity chromatography
- purified by immunoaffinity chromatography
- Concentration:
- 20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
- 20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
- Storage buffer:
- Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice