Anti-LYN Rabbit Polyclonal Antibody
Supplier: Biorbyt
About this item
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200
Type: Primary
Antigen: LYN (v-yes-1 Yamaguchi sarcoma viral related oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
2 Options Available Below
Product Details & Documents
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200
Type: Primary
Antigen: LYN (v-yes-1 Yamaguchi sarcoma viral related oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
orb106346
Datasheet
Specifications for Product Family
Specifications
- Catalog No:
- BIRBORB106346-100
- BIRBORB106346-20
- Antigen symbol:
- LYN
- LYN
- Antigen name:
- v-yes-1 Yamaguchi sarcoma viral related oncogene homolog
- v-yes-1 Yamaguchi sarcoma viral related oncogene homolog
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human|Rat|Mouse
- Human|Rat|Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- A synthesized peptide derived from human Lyn
- A synthesized peptide derived from human Lyn
- Molecular weight:
- 53, 56 kDa
- 53, 56 kDa
- Purification:
- purified by immunoaffinity chromatography
- purified by immunoaffinity chromatography
- Concentration:
- 20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
- 20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
- Storage buffer:
- Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.BIRBORB106346-100Antigen symbolLYNAntigen namev-yes-1 Yamaguchi sarcoma viral related oncogene homologAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHuman|Rat|MouseHostRabbitIsotypeIgGWestern blotYesImmunoChemistryYesImmunogenA synthesized peptide derived from human LynMolecular weight53, 56 kDaPurificationpurified by immunoaffinity chromatographyConcentration20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ulStorage bufferPhosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.Storage temperatureCoolShipping temperatureIce - Specifications:
More
Specifications
Cat. No.BIRBORB106346-20Antigen symbolLYNAntigen namev-yes-1 Yamaguchi sarcoma viral related oncogene homologAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHuman|Rat|MouseHostRabbitIsotypeIgGWestern blotYesImmunoChemistryYesImmunogenA synthesized peptide derived from human LynMolecular weight53, 56 kDaPurificationpurified by immunoaffinity chromatographyConcentration20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ulStorage bufferPhosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.Storage temperatureCoolShipping temperatureIce



