Order Entry
Export
ContactUsLinkComponent
Anti-PPM1B Rabbit Polyclonal Antibody
Anti-PPM1B Rabbit Polyclonal Antibody
Supplier:  Biorbyt
Certificates
undefined
Anti-PPM1B Rabbit Polyclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB101804-100
  • BIRBORB101804-200
  • Antigen symbol:
  • PPM1B
  • PPM1B
  • Antigen name:
  • protein phosphatase, Mg2+/Mn2+ dependent, 1B
  • protein phosphatase, Mg2+/Mn2+ dependent, 1B
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • liquid
  • liquid
  • Molecular weight:
  • 21 kDa
  • 21 kDa
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Concentration:
  • 0,5 mg/ml
  • 0,5 mg/ml
  • Storage buffer:
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information