Anti-NPY1 36 Rabbit Monoclonal Antibody
Catalog # BIRBORB48875-200
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:NPY1 36
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Peptide
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: NPY1 36
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Mouse)
Reactivity: Human