Anti-FABP2 Rabbit polyclonal antibody
Catalog # BIRBORB48372-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Fatty Acid Binding Protein 2, Intestinal
- Antigen symbol:FABP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: FABP2 (fatty acid binding protein 2, intestinal)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat