Order Entry
Export
ContactUsLinkComponent
Anti-CBS Rabbit Polyclonal Antibody
Anti-CBS Rabbit Polyclonal Antibody
Catalog # BIRBORB48303-100
Supplier:  Biorbyt
undefined
Anti-CBS Rabbit Polyclonal Antibody
Catalog # BIRBORB48303-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    cystathionine-beta-synthase
  • Antigen symbol:
    CBS
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: CBS (cystathionine-beta-synthase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat