Anti-SYK Mouse Monoclonal Antibody
Catalog # BIRBORB44501-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Spleen Tyrosine Kinase
- Antigen symbol:SYK
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: SYK (spleen tyrosine kinase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat