Anti-NOVA1 Rabbit Polyclonal Antibody
Catalog # BIRBORB40945-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Neuro-oncological ventral antigen 1
- Antigen symbol:NOVA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Amino acid number:315 to 507
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:0,92 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 315 and 507 of NOVA1.
- Purification:purified by antigen affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200
Type: Primary
Antigen: NOVA1 (neuro-oncological ventral antigen 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human