Anti-HPV (Typ 16 u. 18, E6) Mouse Monoclonal Antibody
Catalog # BIRBORB27224-3
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:HPV (Typ 16 u. 18, E6)
- Antigen symbol:HPV 16/18/E6
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Storage temperature:Cool
- Concentration:neat
- Shipping temperature:Ice
- Size:3 ml
- Pk:3 mL
Specifications
About this item
Type: Primary
Antigen: HPV (Typ 16 u. 18, E6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: