Order Entry
Export
ContactUsLinkComponent
 
Anti-Testosterone 19 Rabbit Polyclonal Antibody
  BIRBORB26787-5000
 :  Biorbyt
Anti-Testosterone 19 Rabbit Polyclonal Antibody
  BIRBORB26787-5000
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    Testosterone 19
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Concentration:
    Titer: 750,000 Tests/ml (125I RIA and 68% B/TC)
  • Shipping temperature:
    Ice
  • Immunogen:
    Testosterone-19-HSA
  • Size:
    5000 test
  • Pk:
    5.000 Tests

 

 

Type: Primary
Antigen: Testosterone 19
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human