Anti-Resistin (33-90) Rabbit Polyclonal Antibody
Catalog # BIRBORB25208-20
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Resistin (33-90)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Form:lyophilized
- Amino acid number:33 to 90
- Sequence:CQSVTSRGDLATCPRGFAVTGCTCGSACGSWDVRAETTCHCQCAGMDWTGARCCRVQP
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic Human Resistin peptide (a.a. 33-90) bTG conjugated (CQSVTSRGDLATCPRGFAVTGCTCGSACGSWDVRAETTCHCQCAGMDWTGARCCRVQP)
- Size:20 µl
- Pk:20 µl
Specifications
About this item
Type: Primary
Antigen: Resistin (33-90)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: