Anti-17-alpha-Hydroxyprogesterone Mouse Monoclonal Antibody
Catalog # BIRBORB24360-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:17-alpha-Hydroxyprogesterone
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG3
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:2,6 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Conjugate of 17-alpha-hydroxyprogesterone with BSA.
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: 17-alpha-Hydroxyprogesterone
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: