Anti-RSV, 33 + 190kDa Mouse Monoclonal Antibody
Catalog # BIRBORB23514-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:RSV, 33 + 190kDa
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Western blot:Yes
- Storage buffer:0,15M PBS, pH 7.0-7.5
- Storage temperature:Cool
- Concentration:1 mg/ml (OD280, E(0,1%)=1,36)
- Shipping temperature:Ice
- Immunogen:Respiratory Syncytial Virus
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: RSV, 33 + 190kDa
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: