Anti-AnthraxLF Rabbit Polyclonal Antibody
BIRBORB22729-200
: Biorbyt
- Antibody type:Primary
- Antigen name:Anthrax Lethal Factor
- Antigen symbol:AnthraxLF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 47-65 of the Anthrax LF protein (~90 kD).
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: AnthraxLF
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Virus