Order Entry
Export
ContactUsLinkComponent
Anti-SARS Spike Mouse Monoclonal Antibody
Anti-SARS Spike Mouse Monoclonal Antibody
Supplier:  Biorbyt
Certificates
Anti-SARS Spike Mouse Monoclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB22727-100
  • BIRBORB22727-200
  • Antigen symbol:
  • SARS Spike
  • SARS Spike
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Virus
  • Virus
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • The antibody was developed using synthetic peptide from the putative SARS Spike glycoprotein(Genbank accession no.NP_828851)corresponding to amino acids 19-35.
  • Amino acid number:
  • 19 to 35
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications