Anti-IgA/IgG1/IgG2a/IgG2b/IgG2c/IgM Goat Polyclonal Antibody
Catalog # BIRBORB21415-10
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen symbol:IgA/IgG1/IgG2a/IgG2b/IgG2c/IgM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Highly purified IgG and pools of homogenous IgA and IgM isolated from rat serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:10 mg
- Pk:10 mg
Specifications
About this item
Type: Secondary
Antigen: Rat IgG1 IgG2A IgG2B IgG2C IgA IgM
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: