Anti-TMEM57 Rabbit Polyclonal Antibody
Catalog # BIRBORB13155-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:transmembrane Protein 57
- Antigen symbol:TMEM57
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Storage buffer:TBS (pH 7,4), 0,5% BSA, 25% Glycerol. Preservative: 0,05% Sodium Azide
- Molecular weight:40 and 60 kDa
- Storage temperature:Cool
- Concentration:2 mg/ml
- Shipping temperature:Ice
- Immunogen:This antibody is produced by immunizing rabbits with a synthetic peptide (KLH-coupled) corresponding to TMEM57.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Western Blot: 1:2000-5000
Type: Primary
Antigen: TMEM57 (transmembrane protein 57)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: