Anti-STIP1 Rabbit Polyclonal Antibody
Supplier: Biorbyt
About this item
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: STIP1 (stress-induced phosphoprotein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
2 Options Available Below
Product Details & Documents
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: STIP1 (stress-induced phosphoprotein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Documents
orb1493
Datasheet
Specifications for Product Family
Specifications
- Catalog No:
- BIRBORB1493-100
- BIRBORB1493-200
- Antigen symbol:
- STIP1
- STIP1
- Antigen name:
- Stress-induced phosphoprotein 1
- Stress-induced phosphoprotein 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide
- Synthetic peptide
- Form:
- liquid
- liquid
- Purification:
- purified by Protein A chromatography
- purified by Protein A chromatography
- Concentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Storage buffer:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.BIRBORB1493-100Antigen symbolSTIP1Antigen nameStress-induced phosphoprotein 1Antibody typePrimaryClonalityPolyclonalConjugationUnconjugatedHostRabbitIsotypeIgGWestern blotYesELISAYesImmunoChemistryYesImmunogenSynthetic peptideFormliquidPurificationpurified by Protein A chromatographyConcentration0,5 mg/mlStorage buffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolStorage temperatureCoolShipping temperatureIce - Specifications:
More
Specifications
Cat. No.BIRBORB1493-200Antigen symbolSTIP1Antigen nameStress-induced phosphoprotein 1Antibody typePrimaryClonalityPolyclonalConjugationUnconjugatedHostRabbitIsotypeIgGWestern blotYesELISAYesImmunoChemistryYesImmunogenSynthetic peptideFormliquidPurificationpurified by Protein A chromatographyConcentration0,5 mg/mlStorage buffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolStorage temperatureCoolShipping temperatureIce



