Anti-TRIM9 Mouse Polyclonal Antibody
ABNOH00114088-A01
: Abnova
- Antibody type:Primary
- Antigen name:Tripartite Motif Containing 9
- Antigen symbol:TRIM9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:114088
- Antigen synonyms:SPRING|RNF91
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: TRIM9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human