Order Entry
Export
ContactUsLinkComponent
Anti-TOE1 Mouse Monoclonal Antibody [clone: 2D7]
Anti-TOE1 Mouse Monoclonal Antibody [clone: 2D7]
Catalog # ABNOH00114034-M04
Supplier:  Abnova
undefined
Anti-TOE1 Mouse Monoclonal Antibody [clone: 2D7]
Catalog # ABNOH00114034-M04
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Target of EGR1, member 1
  • Antigen symbol:
    TOE1
  • Clonality:
    Monoclonal
  • Clone:
    2D7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    114034
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: TOE1
Clonality: Monoclonal
Clone: 2D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human