Anti-NAPRT Rabbit Polyclonal Antibody
ABNOH00093100-D01
: Abnova
- Antibody type:Primary
- Antigen name:Nicotinate phosphoribosyltransferase domain containing 1
- Antigen symbol:NAPRT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:93100
- Antigen synonyms:NAPRT1|PP3856
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: NAPRT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human