Anti-KLC4 Mouse Polyclonal Antibody
Catalog # ABNOH00089953-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:kinesin light chain 4
- Antigen symbol:KLC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:89953
- Antigen synonyms:bA387M24.3|KNSL8
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: KLC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat