Anti-CARF Mouse Polyclonal Antibody
50 µg
ABNOH00079800-B01P
: Abnova
Type: Primary
Antigen: CARF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Size:50 µg
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- ImmunoFluorescence:Yes
- Storage temperature:Cool
- Host:Mouse
- Cross adsorption:No
- Shipping temperature:Ice
- Antigen name:calcium responsive transcription factor
- Cat. No.:ABNOH00079800-B01P
- Reactivity:Human
- Gene ID:79800
- Antigen synonyms:ALS2CR8|NYD-SP24
- Conjugation:Unconjugated
- Antigen symbol:CARF



