Anti-SMC6 Mouse Monoclonal Antibody [clone: 2E6]
ABNOH00079677-M01
: Abnova
- Antibody type:Primary
- Antigen name:Structural maintenance of chromosomes 6
- Antigen symbol:SMC6
- Clonality:Monoclonal
- Clone:2E6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:79677
- Antigen synonyms:SMC6 protein|SMC6 structural maintenance of chromosomes 6 like 1|SMC6L1|SMC 6|SMC6|Structural maintenance of chromosomes protein 6.|Structural maintenance of chromosomes 6 like 1|Structural maintenance of chromosomes 6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SMC6
Clonality: Monoclonal
Clone: 2E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human