Anti-GLB1L Mouse Polyclonal Antibody
ABNOH00079411-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Galactosidase, beta 1-like
- Antigen symbol:GLB1L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:79411
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: GLB1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human