Anti-TRIM49 Mouse Monoclonal Antibody [clone: 2A3]
Catalog # ABNOH00057093-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tripartite Motif Containing 49
- Antigen symbol:TRIM49
- Clonality:Monoclonal
- Clone:2A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:57093
- Antigen synonyms:TRIM49L2|RNF18|TRIM49A
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TRIM49
Clonality: Monoclonal
Clone: 2A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human