Anti-ARMCX3 Mouse Polyclonal Antibody
Catalog # ABNOH00051566-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Armadillo Repeat Containing, X-linked 3
- Antigen symbol:ARMCX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:51566
- Antigen synonyms:Arm protein lost in epithelial cancers|dJ545K15.2|GASP6|ARM protein lost in epithelial cancers on chromosome X 3|Armadillo repeat containing X linked protein 3|X chromosome|ALEX3|3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: ARMCX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human