Order Entry
Export
ContactUsLinkComponent
Anti-MBIP Mouse Monoclonal Antibody [clone: 2E2]
Anti-MBIP Mouse Monoclonal Antibody [clone: 2E2]
Catalog # ABNOH00051562-M05
Supplier:  Abnova
undefined
Anti-MBIP Mouse Monoclonal Antibody [clone: 2E2]
Catalog # ABNOH00051562-M05
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MAP3K12-binding inhibitory protein 1
  • Antigen symbol:
    MBIP
  • Clonality:
    Monoclonal
  • Clone:
    2E2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Gene ID:
    51562
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: MBIP
Clonality: Monoclonal
Clone: 2E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human