Anti-TMOD3 Mouse Monoclonal Antibody [clone: 1E1]
Catalog # ABNOH00029766-M10
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tropomodulin 3
- Antigen symbol:TMOD3
- Clonality:Monoclonal
- Clone:1E1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:29766
- Antigen synonyms:UTMOD
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TMOD3
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human