Anti-DUX3 Mouse Monoclonal Antibody [clone: 3D2]
ABNOH00026582-M01
: Abnova
- Antibody type:Primary
- Antigen name:Double homeobox 3
- Antigen symbol:DUX3
- Clonality:Monoclonal
- Clone:3D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:26582
- Antigen synonyms:double homeobox 3|Putative double homeobox protein 3
- Storage buffer:In 1X PBS, pH 7,4
- Sequence:MALLTALDDTLPEEAQGPGRRMILLSTPSQSDALRACFERNLYPGIATKEQLAQGIDIPEPRVQIWFQNERSCQLRQHRRQSRPWPGRRDPQKGR
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Shipping temperature:Ice
- Immunogen:DUX3 (NP_036280, 1 a.a. ~ 95 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 kDa.
- Size:100 µg
- Pk:100 µg
Mouse monoclonal antibody raised against a partial recombinant DUX3.
Recommended Dilutons: Western Blot (1 to 5 µg/ml).
Type: Primary
Antigen: DUX3
Clonality: Monoclonal
Clone: 3D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human