Anti-IKZF3 Mouse Monoclonal Antibody [clone: 1D7]
Catalog # ABNOH00022806-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ikaros family Zinc Finger Protein 3
- Antigen symbol:IKZF3
- Clonality:Monoclonal
- Clone:1D7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:22806
- Antigen synonyms:AIO|AIOLOS|ZNFN1A3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: IKZF3
Clonality: Monoclonal
Clone: 1D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human