Anti-GABARAPL2 Mouse Polyclonal Antibody
Catalog # ABNOH00011345-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Gamma-aminobutyric acid receptor-associated protein-like 2
- Antigen symbol:GABARAPL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11345
- Antigen synonyms:GEF2|ATG8|ATG8C|GATE16|GEF-2|GATE-16
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GABARAPL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human