Anti-DNAJB4 Mouse Monoclonal Antibody [clone: S13]
Catalog # ABNOH00011080-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:DnaJ (Hsp40) homolog, subfamily B, Member 4
- Antigen symbol:DNAJB4
- Clonality:Monoclonal
- Clone:S13
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11080
- Antigen synonyms:HLJ1|HSP40 homolog|DnaJ like heat shock protein 40|HLJ 1|Heat shock protein 40 homolog|Heat shock 40 kDa protein 1 homolog|DNAJB 4|DnaJ (Hsp40) homolog subfamily B member 4|DnaJ homolog subfamily B member 4|DNAJW|DjB4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: DNAJB4
Clonality: Monoclonal
Clone: S13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human