Order Entry
Export
ContactUsLinkComponent
Anti-ZNF268 Mouse Monoclonal Antibody [clone: 3B4]
Anti-ZNF268 Mouse Monoclonal Antibody [clone: 3B4]
  ABNOH00010795-M02
 :  Abnova
undefined
Anti-ZNF268 Mouse Monoclonal Antibody [clone: 3B4]
  ABNOH00010795-M02
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Zinc Finger Protein 268
  • Antigen symbol:
    ZNF268
  • Clonality:
    Monoclonal
  • Clone:
    3B4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Gene ID:
    10795
  • Antigen synonyms:
    HZF3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: ZNF268
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human