Order Entry
Export
ContactUsLinkComponent
Anti-TBR1 Mouse Monoclonal Antibody [clone: 3F6]
Anti-TBR1 Mouse Monoclonal Antibody [clone: 3F6]
Catalog # ABNOH00010716-M01
Supplier:  Abnova
undefined
Anti-TBR1 Mouse Monoclonal Antibody [clone: 3F6]
Catalog # ABNOH00010716-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    T-box, brain, 1
  • Antigen symbol:
    TBR1
  • Clonality:
    Monoclonal
  • Clone:
    3F6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    10716
  • Antigen synonyms:
    it defines dferent regions that give rise to the palecortex|limbic cortex and neocortex 1-3. TBR-1|TBR-1|where|during embryogenesis|A novel murine and human gene|encodes a putative transcription factor related to the Brachyrury T gene that is expressed only in postmitotic cells. T-brain-1 TBR-1 mRNA is largely restricted to the cerebral cortex|Pax-6 and Emx-1 are expressed in the mouse and chicken pallium. The pallio-subpallial boundary lies at the interface between the TBR-1 and Dlx-2 expression domains. .
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: TBR1
Clonality: Monoclonal
Clone: 3F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human