Order Entry
Export
ContactUsLinkComponent
Anti-USP39 Mouse Polyclonal Antibody
Anti-USP39 Mouse Polyclonal Antibody
Catalog # ABNOH00010713-B01P
Supplier:  Abnova
undefined
Anti-USP39 Mouse Polyclonal Antibody
Catalog # ABNOH00010713-B01P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitin specific peptidase 39
  • Antigen symbol:
    USP39
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    10713
  • Antigen synonyms:
    65K|SAD1|CGI-21|HSPC332|SNRNP65
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µg

Specifications

About this item

Type: Primary
Antigen: USP39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human