Anti-CORIN Mouse Monoclonal Antibody [clone: 5B6]
Catalog # ABNOH00010699-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Corin, serine peptidase
- Antigen symbol:CORIN
- Clonality:Monoclonal
- Clone:5B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10699
- Antigen synonyms:CRN|TMPRSS10|Lrp4|ATC2|PEE5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CORIN
Clonality: Monoclonal
Clone: 5B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human