Anti-BCAS2 Mouse Polyclonal Antibody
ABNOH00010286-A01
: Abnova
- Antibody type:Primary
- Antigen name:Breast carcinoma-amplified sequence 2
- Antigen symbol:BCAS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10286
- Antigen synonyms:Breast carcinoma-amplified sequence 2|BCAS2|Pre-mRNA-splicing factor SPF27|SPF27|DAM1|Snt309
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: BCAS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human