Order Entry
Export
ContactUsLinkComponent
Anti-UBE4B Mouse Polyclonal Antibody
Anti-UBE4B Mouse Polyclonal Antibody
Catalog # ABNOH00010277-A01
Supplier:  Abnova
Anti-UBE4B Mouse Polyclonal Antibody
Catalog # ABNOH00010277-A01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitination Factor E4B
  • Antigen symbol:
    UBE4B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Gene ID:
    10277
  • Antigen synonyms:
    Ubiquitin-fusion degradation protein 2|KIAA0684|Homozygously deleted in neuroblastoma-1|HDNB1|Ubiquitin conjugation factor E4 B|UFD2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: UBE4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human