Order Entry
Export
ContactUsLinkComponent
 
Anti-KMT2B Mouse Monoclonal Antibody [clone: 1F6]
  ABNOH00009757-M01
 :  Abnova
undefined
Anti-KMT2B Mouse Monoclonal Antibody [clone: 1F6]
  ABNOH00009757-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Lysine (K)-Specific Methyltransferase 2B
  • Antigen symbol:
    KMT2B
  • Clonality:
    Monoclonal
  • Clone:
    1F6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    9757
  • Antigen synonyms:
    MLL4|CXXC10|TRX2|WBP7|MLL2|WBP-7|HRX2|MLL1B
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: KMT2B
Clonality: Monoclonal
Clone: 1F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human