Anti-NAE1 Mouse Monoclonal Antibody [clone: 4E8-H3]
Catalog # ABNOH00008883-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:NEDD8 activating enzyme E1 subunit 1
- Antigen symbol:NAE1
- Clonality:Monoclonal
- Clone:4E8-H3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8883
- Antigen synonyms:APPBP1|A-116A10.1|ula-1|HPP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: NAE1
Clonality: Monoclonal
Clone: 4E8-H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human