Anti-NR2F2 Mouse Monoclonal Antibody [clone: 2G12]
Catalog # ABNOH00007026-M15A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Receptor Subfamily 2, Group F, Member 2
- Antigen symbol:NR2F2
- Clonality:Monoclonal
- Clone:2G12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7026
- Antigen synonyms:ARP1, CHTD4, COUPTFB, COUPTFII, NF-E3, NR2F1, SVP40, TFCOUP2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: NR2F2
Clonality: Monoclonal
Clone: 2G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human